Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nqo2
Entrez Gene ID
291084
Gene Name
NAD(P)H dehydrogenase, quinone 2
Gene Aliases
-
Chromosome Location
Chr.17: 32131847 - 32158558 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006253868.2 XP_006253930.1 1 - 2 121 74

View Full Product Details