The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017036.2 | NP_058732.2 | 3 - 4 | 335 | 84 |
GenBank mRNA | BC097332.1 | - | 3 - 4 | 335 | 84 |
FQ228460.1 | - | 3 - 4 | 370 | 84 | |
M13750.1 | - | 3 - 4 | 318 | 84 |