The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106128.1 | NP_001099598.1 | 1 - 2 | 233 | 60 |
XM_006253800.2 | XP_006253862.1 | 2 - 3 | 405 | 60 |