Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Foxc1
Entrez Gene ID
364706
Gene Name
forkhead box C1
Gene Aliases
-
Chromosome Location
Chr.17: 33947501 - 33951484 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134338.1 NP_599165.1 1 - 1 3290 138

View Full Product Details