Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bmp6
Entrez Gene ID
25644
Gene Name
bone morphogenetic protein 6
Gene Aliases
VGR
Chromosome Location
Chr.17: 26955142 - 27112820 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013107.1 NP_037239.1 3 - 4 985 116
GenBank mRNA AY184240.1 - 3 - 4 985 116
X58830.1 - - 88 116

View Full Product Details