The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105751.1 | NP_001099221.1 | 8 - 9 | 6569 | 62 |
GenBank mRNA | X54250.1 | - | 3 - 4 | 381 | 62 |