Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Sptlc1
- Entrez Gene ID
-
361213
- Gene Name
- serine palmitoyltransferase, long chain base subunit 1
- Gene Aliases
-
Lcb1, RGD1306617, Spt1
- Chromosome Location
-
Chr.17: 11856525 - 11895566 on Build Rnor_6.0
- Gene Symbol
- Cyp3a23/3a1
- Entrez Gene ID
-
25642
- Gene Name
- cytochrome P450, family 3, subfamily a, polypeptide 23/polypeptide 1
- Gene Aliases
-
CYP, CYP3A23, Cyp3a1, Cyp3a3, RL33, cDEX
- Chromosome Location
-
Chr.12: 11053888 - 11082742 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
FQ218135.1 |
- |
- |
275 |
70 |
View Full Product Details