The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006966.1 | NP_001006967.1 | 9 - 10 | 1053 | 90 |
GenBank mRNA | BC083764.1 | - | 9 - 10 | 1053 | 90 |