Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pak1ip1
Entrez Gene ID
361232
Gene Name
PAK1 interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.17: 21579987 - 21591064 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037356.1 NP_001032433.1 7 - 8 801 135
GenBank mRNA BC079236.1 - 7 - 8 781 135
BC089822.1 - 8 - 9 862 135
BC107922.1 - 7 - 8 801 135

View Full Product Details