Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zdhhc12
Entrez Gene ID
366014
Gene Name
zinc finger, DHHC-type containing 12
Gene Aliases
-
Chromosome Location
Chr.3: 8656397 - 8659102 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013239.1 NP_001013257.1 2 - 3 254 75
GenBank mRNA AY886528.1 - 2 - 3 254 75
BC076393.1 - 2 - 3 254 75

View Full Product Details