Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nelfb
Entrez Gene ID
311796
Gene Name
negative elongation factor complex member B
Gene Aliases
Cobra1, RGD1307832
Chromosome Location
Chr.3: 2414451 - 2431148 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107817.2 NP_001101287.2 6 - 7 1112 61
GenBank mRNA BC160853.1 - 5 - 6 1234 61

View Full Product Details