Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Brd3
Entrez Gene ID
362092
Gene Name
bromodomain containing 3
Gene Aliases
-
Chromosome Location
Chr.3: 5995204 - 6054470 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006233825.2 XP_006233887.1 8 - 9 1610 100
XM_006233826.2 XP_006233888.1 8 - 9 1636 100
GenBank mRNA BC168949.1 - 8 - 9 1555 100

View Full Product Details