Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrrf
Entrez Gene ID
311903
Gene Name
mitochondrial ribosome recycling factor
Gene Aliases
-
Chromosome Location
Chr.3: 15463919 - 15520621 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008354.1 NP_001008355.1 3 - 4 384 110
XM_006234068.1 XP_006234130.1 3 - 4 418 110
GenBank mRNA BC085779.1 - 3 - 4 384 110

View Full Product Details