Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Osgin2
Entrez Gene ID
313085
Gene Name
oxidative stress induced growth inhibitor family member 2
Gene Aliases
RGD1306323
Chromosome Location
Chr.5: 29663131 - 29682057 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006237919.2 XP_006237981.1 4 - 5 842 110
XM_232798.8 XP_232798.4 4 - 5 672 110

View Full Product Details