Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmod1
Entrez Gene ID
25566
Gene Name
tropomodulin 1
Gene Aliases
E-Tmod, Tmod
Chromosome Location
Chr.5: 61635611 - 61712052 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013044.2 NP_037176.2 6 - 7 708 126
GenBank mRNA BC072521.1 - 6 - 7 708 126
U59241.1 - 5 - 6 664 126

View Full Product Details