Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hint2
Entrez Gene ID
313491
Gene Name
histidine triad nucleotide binding protein 2
Gene Aliases
-
Chromosome Location
Chr.5: 59150344 - 59152599 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107955.2 NP_001101425.2 2 - 3 222 108
GenBank mRNA BI282127.1 - 3 - 4 108

View Full Product Details