The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106658.2 | NP_001100128.1 | 8 - 9 | 1228 | 82 |
GenBank mRNA | BC161967.1 | - | 8 - 9 | 1228 | 82 |