Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb5
Entrez Gene ID
298084
Gene Name
zinc finger and BTB domain containing 5
Gene Aliases
-
Chromosome Location
Chr.5: 60538926 - 60561491 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106657.1 NP_001100127.1 1 - 2 207 107

View Full Product Details