The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080407.1 | NP_536332.1 | 7 - 8 | 1336 | 104 |
XM_006244951.2 | XP_006245013.1 | 8 - 9 | 1822 | 104 | |
XM_006244952.2 | XP_006245014.1 | 8 - 9 | 1822 | 104 |