Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tyw5
Entrez Gene ID
301419
Gene Name
tRNA-yW synthesizing protein 5
Gene Aliases
C2orf60, RGD1311269
Chromosome Location
Chr.9: 64077841 - 64096265 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033170.1 2 - 3 480 146
GenBank mRNA BC158619.1 - 2 - 3 480 146

View Full Product Details