Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mdh1b
Entrez Gene ID
316444
Gene Name
malate dehydrogenase 1B, NAD (soluble)
Gene Aliases
-
Chromosome Location
Chr.9: 70404093 - 70450170 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108221.1 NP_001101691.1 4 - 5 467 92

View Full Product Details