The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013112.1 | NP_001013130.1 | 9 - 10 | 852 | 151 |
XM_006244895.1 | XP_006244957.1 | 9 - 10 | 938 | 151 | |
GenBank mRNA | BC083737.1 | - | 9 - 10 | 852 | 151 |