Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tgfbrap1
Entrez Gene ID
301373
Gene Name
transforming growth factor, beta receptor associated protein 1
Gene Aliases
-
Chromosome Location
Chr.9: 49852560 - 49902943 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106907.1 NP_001100377.1 10 - 11 2365 74
XM_006244788.2 XP_006244850.1 9 - 10 1959 74
XM_006244789.2 XP_006244851.1 9 - 10 1912 74
XM_006244790.2 XP_006244852.1 9 - 10 1918 74
XM_006244791.2 XP_006244853.1 9 - 10 1938 74
XM_006244792.2 XP_006244854.1 9 - 10 1856 74
XM_006244793.2 XP_006244855.1 9 - 10 2681 74
XM_008767021.1 XP_008765243.1 9 - 10 1997 74

View Full Product Details