Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prss40
Entrez Gene ID
316318
Gene Name
protease, serine, 40
Gene Aliases
RGD1561330, Tesp2
Chromosome Location
Chr.9: 41061116 - 41069929 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108209.1 NP_001101679.1 3 - 4 637 140

View Full Product Details