The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106913.2 | NP_001100383.2 | 12 - 13 | 1443 | 112 |
XM_008767083.1 | XP_008765305.1 | 11 - 12 | 1372 | 112 | |
GenBank mRNA | AA923848.1 | - | 1 - 2 | 112 | |
BC081942.1 | - | 10 - 11 | 2702 | 112 | |
BC097451.1 | - | 10 - 11 | 2659 | 112 | |
FQ232068.1 | - | 12 - 13 | 1453 | 112 |