The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014100.1 | NP_001014122.1 | 2 - 3 | 729 | 86 |
XM_008767025.1 | XP_008765247.1 | 3 - 4 | 1025 | 86 | |
XM_008767026.1 | XP_008765248.1 | 3 - 4 | 803 | 86 | |
GenBank mRNA | BC088198.1 | - | 2 - 3 | 729 | 86 |
FQ227363.1 | - | 2 - 3 | 794 | 86 |