Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mfsd6
Entrez Gene ID
301388
Gene Name
major facilitator superfamily domain containing 6
Gene Aliases
RGD1562317
Chromosome Location
Chr.9: 53625579 - 53710820 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106911.1 NP_001100381.1 3 - 4 1993 147
XM_006244897.2 XP_006244959.1 2 - 3 3404 147
XM_006244898.2 XP_006244960.1 2 - 3 3030 147

View Full Product Details