Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cfc1
Entrez Gene ID
501121
Gene Name
cripto, FRL-1, cryptic family 1
Gene Aliases
RGD1562188
Chromosome Location
Chr.9: 41045352 - 41051979 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109304.1 NP_001102774.1 - 705 57

View Full Product Details