The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031510.1 | NP_113698.1 | 1 - 2 | 145 | 121 |
XM_006245049.2 | XP_006245111.1 | 2 - 3 | 203 | 121 | |
XM_008767076.1 | XP_008765298.1 | 2 - 3 | 235 | 121 | |
GenBank mRNA | FQ214242.1 | - | 2 - 3 | 226 | 121 |
L35317.1 | - | 1 - 2 | 145 | 121 |