The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031348.2 | NP_112638.2 | 9 - 10 | 855 | 88 |
GenBank mRNA | AB026057.2 | - | 9 - 10 | 766 | 88 |
BC088128.1 | - | 9 - 10 | 855 | 88 |