Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Uap1l1
Entrez Gene ID
296560
Gene Name
UDP-N-acteylglucosamine pyrophosphorylase 1-like 1
Gene Aliases
-
Chromosome Location
Chr.3: 2579250 - 2584523 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134516.1 NP_001127988.1 2 - 3 569 97
GenBank mRNA BC168670.1 - 2 - 3 569 97

View Full Product Details