The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100171.1 | NP_001093641.1 | 3 - 4 | 368 | 64 |
XM_006233600.2 | XP_006233662.1 | 3 - 4 | 655 | 64 |