Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St6galnac4
Entrez Gene ID
407764
Gene Name
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 4
Gene Aliases
siat7D
Chromosome Location
Chr.3: 11607203 - 11619595 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003749452.3 XP_003749500.2 5 - 6 991 76
XM_006234012.2 XP_006234074.1 4 - 5 1153 76
XM_008761787.1 XP_008760009.1 3 - 4 967 76
GenBank mRNA AJ646871.1 - 3 - 4 722 76

View Full Product Details