Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccbl1
Entrez Gene ID
311844
Gene Name
cysteine conjugate-beta lyase, cytoplasmic
Gene Aliases
Gtk, Kat1, KatI
Chromosome Location
Chr.3: 8752289 - 8785615 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013164.3 NP_001013182.3 1 - 2 96 87
XM_008761658.1 XP_008759880.1 2 - 3 779 87
XM_008761659.1 XP_008759881.1 2 - 3 783 87
GenBank mRNA Z49696.1 - 2 - 3 245 87

View Full Product Details