Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Retnlb
Entrez Gene ID
498074
Gene Name
resistin like beta
Gene Aliases
-
Chromosome Location
Chr.11: 54565471 - 54567357 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024281.1 NP_001019452.1 3 - 4 244 68
XM_008768646.1 XP_008766868.1 2 - 3 313 68
GenBank mRNA AY641429.1 - 3 - 4 244 68

View Full Product Details