Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dtx3l
Entrez Gene ID
498089
Gene Name
deltex 3 like, E3 ubiquitin ligase
Gene Aliases
RGD1565144
Chromosome Location
Chr.11: 67758011 - 67767622 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109053.1 NP_001102523.1 2 - 3 556 94
XM_006248414.2 XP_006248476.1 2 - 3 471 94

View Full Product Details