Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csta
Entrez Gene ID
288067
Gene Name
cystatin A (stefin A)
Gene Aliases
-
Chromosome Location
Chr.11: 67555792 - 67566980 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105876.1 NP_001099346.1 1 - 2 104 68

View Full Product Details