Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rngtt
Entrez Gene ID
313131
Gene Name
RNA guanylyltransferase and 5'-phosphatase
Gene Aliases
-
Chromosome Location
Chr.5: 48561325 - 48779214 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107923.2 NP_001101393.1 3 - 4 392 140
XM_006237990.2 XP_006238052.1 3 - 4 406 140
XM_006237991.2 XP_006238053.1 3 - 4 378 140
GenBank mRNA BC168990.1 - 3 - 4 388 140

View Full Product Details