Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ube2j1
Entrez Gene ID
297961
Gene Name
ubiquitin-conjugating enzyme E2, J1
Gene Aliases
-
Chromosome Location
Chr.5: 48274387 - 48292699 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106642.1 NP_001100112.1 5 - 6 457 83
XM_008763612.1 XP_008761834.1 5 - 6 555 83
XM_008763613.1 XP_008761835.1 5 - 6 555 83

View Full Product Details