Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d2
Entrez Gene ID
313234
Gene Name
TBC1 domain family, member 2
Gene Aliases
-
Chromosome Location
Chr.5: 62213001 - 62261468 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238056.2 XP_006238118.1 1 - 2 1800 85
GenBank mRNA BC168982.1 - 1 - 2 405 85

View Full Product Details