Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnai1
Entrez Gene ID
500442
Gene Name
dynein, axonemal, intermediate chain 1
Gene Aliases
Dnaic1
Chromosome Location
Chr.5: 57947796 - 58017985 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024342.1 NP_001019513.1 2 - 3 172 128
GenBank mRNA BC083662.1 - 2 - 3 172 128

View Full Product Details