Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tpm2
Entrez Gene ID
500450
Gene Name
tropomyosin 2, beta
Gene Aliases
-
Chromosome Location
Chr.5: 59016616 - 59025971 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024345.2 NP_001019516.1 6 - 7 806 105
XM_008763681.1 XP_008761903.1 6 - 7 1146 105
GenBank mRNA BC090009.1 - 6 - 7 727 105
CF110059.1 - 6 - 7 692 105
FQ217324.1 - 6 - 7 743 105
FQ219788.1 - 6 - 7 703 105
FQ219821.1 - 6 - 7 743 105
FQ220438.1 - 6 - 7 703 105
FQ220606.1 - 6 - 7 743 105
FQ220675.1 - 6 - 7 743 105
FQ229470.1 - 6 - 7 743 105

View Full Product Details