Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Otud6b
Entrez Gene ID
297911
Gene Name
OTU domain containing 6B
Gene Aliases
RGD1310024
Chromosome Location
Chr.5: 28333019 - 28350104 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106639.1 NP_001100109.1 6 - 7 1013 98

View Full Product Details