Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo10
Entrez Gene ID
362511
Gene Name
F-box protein 10
Gene Aliases
-
Chromosome Location
Chr.5: 60589881 - 60636553 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238141.2 XP_006238203.1 2 - 3 942 81
XM_008763689.1 XP_008761911.1 2 - 3 771 81
XM_342829.7 XP_342830.4 - 721 81

View Full Product Details