Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nans
Entrez Gene ID
298071
Gene Name
N-acetylneuraminic acid synthase
Gene Aliases
-
Chromosome Location
Chr.5: 62109352 - 62126492 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106655.2 NP_001100125.2 5 - 6 903 62
GenBank mRNA BC161979.1 - 5 - 6 897 62
BG669394.1 - 1 - 2 260 62

View Full Product Details