Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gne
Entrez Gene ID
114711
Gene Name
glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase
Gene Aliases
Uae1
Chromosome Location
Chr.5: 59512869 - 59553423 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053765.2 NP_446217.1 11 - 12 2275 59
XM_006238006.2 XP_006238068.1 11 - 12 2093 59
XM_006238008.1 XP_006238070.1 11 - 12 2059 59
XM_006238009.2 XP_006238071.1 11 - 12 2436 59
GenBank mRNA BC062011.1 - 11 - 12 2275 59
FQ224587.1 - 10 - 11 1943 59
Y07744.1 - 10 - 11 1983 59

View Full Product Details