Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cpne3
Entrez Gene ID
313087
Gene Name
copine III
Gene Aliases
-
Chromosome Location
Chr.5: 33525307 - 33574608 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107917.2 NP_001101387.2 11 - 12 1068 168
XM_006237932.1 XP_006237994.1 12 - 13 1190 168
GenBank mRNA AM746508.1 - 11 - 12 1068 168

View Full Product Details