Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Themis
Entrez Gene ID
498985
Gene Name
thymocyte selection associated
Gene Aliases
RGD1560849
Chromosome Location
Chr.1: 17279910 - 17378294 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008758640.1 XP_008756862.1 2 - 3 778 120

View Full Product Details