The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025640.1 | NP_001020811.1 | 6 - 7 | 678 | 76 |
XM_006227646.2 | XP_006227708.1 | 7 - 8 | 828 | 76 | |
GenBank mRNA | BC098787.1 | - | 6 - 7 | 678 | 76 |