Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nhsl1
Entrez Gene ID
308631
Gene Name
NHS-like 1
Gene Aliases
RGD1308124
Chromosome Location
Chr.1: 13568486 - 13705360 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002728625.4 XP_002728671.1 1 - 2 2233 118

View Full Product Details